Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163759.1 | NP_001157231.1 | 18 - 19 | 3817 | 63 | |
NM_198942.2 | NP_945180.2 | 17 - 18 | 3658 | 63 | ||
GenBank mRNA | BC026474.1 | - | 5 - 6 | 799 | 63 | |
BC037692.1 | - | 6 - 7 | 894 | 63 | ||
BC062952.1 | - | 17 - 18 | 3658 | 6853031 | 63 | |
BC065169.1 | - | 18 - 19 | 3484 | 6831314 | 63 | |
BC066091.1 | - | 17 - 18 | 3320 | 6841207 | 63 | |
BC079618.1 | - | 17 - 18 | 3938 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |