Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233459_m1
          See other GM1976 GE Assays ›
          Gene Symbol:
          Gm1976
          Gene Name:
          predicted gene 1976
          Gene Aliases:
          100038868
          Chromosome Location:
          Chr.17: 94759942 - 94834799 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233459_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm1976
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045963.1 2 - 3 166 207
          NR_045964.1 2 - 3 166 150
          GenBank mRNA AK082664.1 - 2 - 3 166 207
          AK137318.1 - 2 - 3 167 150

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK082664.1, AK137318.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120205 AK082664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120205 AK137318.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 180151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125215 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103447 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131583 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186480 AK049147.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08895259 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08895316 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08895296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03400277 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120205 AK082664.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120205 AK137318.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112953 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103447 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131583 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03400277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31456191 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453558 - -
          Affymetrix HT MG-430 PM Array Plate 1455715_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700024707.mm.1 NR_045963.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024707.mm.1 NR_045963.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024707.mm.1 NR_045964.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024707.mm.1 NR_045964.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024707.mm.1 TC1700002757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024702.mm.1 NR_045963.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024702.mm.1 NR_045963.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024702.mm.1 TC1700002757.mm.1 MTA TC
          Mouse Expression Set 430 1455715_PM_at - -
          Mouse Expression Set 430 1455715_at - -
          Murine Genome U74v2 Set 166856_f_at - -
          Murine Genome U74v2 Set 166856_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1455715_at - -
          Mouse Genome 430A 2.0 Array 1455715_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104280487 - -
          Illumina mouse-6 v1.0 expression beadchip 104280487 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235668 - -
          Illumina Sentrix mouse-6.v1 bead array 104280487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104280487 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2587638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0