Search
Search

Plekhb2| Gene Symbol: Plekhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145516.2 | NP_663491.1 | 5 - 6 | 445 | 77 | |
| NM_175421.1 | NP_780630.1 | 5 - 6 | 445 | 77 | ||
| GenBank mRNA | AF189817.1 | - | 5 - 6 | 405 | 77 | |
| AK082118.1 | - | 5 - 6 | 398 | 77 | ||
| AK148653.1 | - | 5 - 6 | 428 | 77 | ||
| AK149868.1 | - | 5 - 6 | 405 | 77 | ||
| AK150074.1 | - | 5 - 6 | 405 | 77 | ||
| AK166091.1 | - | 5 - 6 | 413 | 77 | ||
| BC057994.1 | - | 5 - 6 | 413 | 5344583 | 77 | |
| CK625615.1 | - | 5 - 6 | 445 | 77 | ||
Set Membership: |
|