Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234785_m1
          See other GM15663 GE Assays ›
          Gene Symbol:
          Gm15663
          Gene Name:
          predicted gene 15663
          Gene Aliases:
          9330159K06, OTTMUSG00000025047
          Chromosome Location:
          Chr.10: 105574551 - 105583870 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234785_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15663
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038032.1 2 - 3 1323 70
          GenBank mRNA AK034146.1 - 2 - 3 1316 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001254.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22288 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130364 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02209666 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130364 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27416855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08373707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27416861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27416854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27416856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27416860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434818_PM_at - -
          Mouse Genome 430 2.0 Array 1434818_PM_at - -
          Mouse Genome 430 2.0 Array 1434818_at - -
          Affymetrix Mouse Clariom S Array PSR1000009432.mm.1 NR_038032.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009432.mm.1 NR_038032.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009432.mm.1 TC1000001254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009433.mm.1 NR_038032.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009433.mm.1 NR_038032.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009433.mm.1 TC1000001254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009431.mm.1 NR_038032.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009431.mm.1 NR_038032.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009431.mm.1 TC1000001254.mm.1 MTA TC
          Mouse Expression Set 430 1434818_PM_at - -
          Mouse Expression Set 430 1434818_at - -
          Murine Genome U74v2 Set 167984_r_at - -
          Murine Genome U74v2 Set 169444_PM_at - -
          Murine Genome U74v2 Set 169444_at - -
          Murine Genome U74v2 Set 167984_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1434818_at - -
          Affymetrix HT Mouse Genome 430B Array 1434818_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 853730 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline