Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234804_m1
          See other 4930563E18RIK GE Assays ›
          Gene Symbol:
          4930563E18Rik
          Gene Name:
          RIKEN cDNA 4930563E18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 10706860 - 10711832 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234804_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930563E18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045379.1 2 - 3 1391 81
          GenBank mRNA AK016200.1 - 2 - 3 1384 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000083.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2405853 AK016200.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482973 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162958 - -
          Whole Mouse Genome G4122A A_52_P1123245 AK016200.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116575 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130679 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162958 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432031_PM_at - -
          Mouse Genome 430 2.0 Array 1432031_PM_at - -
          Mouse Genome 430 2.0 Array 1432031_at - -
          Affymetrix Mouse Clariom S Array PSR1800000528.mm.1 NR_045379.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000528.mm.1 NR_045379.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000528.mm.1 TC1800000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000530.mm.1 NR_045379.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000530.mm.1 NR_045379.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000530.mm.1 TC1800000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000529.mm.1 NR_045379.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000529.mm.1 NR_045379.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000529.mm.1 TC1800000083.mm.1 MTA TC
          Mouse Expression Set 430 1432031_at - -
          Mouse Expression Set 430 1432031_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432031_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432031_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 514681 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103940725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline