| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639944 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
300434 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547396 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691757 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
123813 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948139 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384184 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
735769 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824232 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283638 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674324 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
247522 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26036 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95666 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
231623 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
423025 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
22659 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3578 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702114 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683066 |
AK003321.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683066 |
AK009696.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683066 |
BC038674.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683066 |
NM_025966.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
772501 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
418260 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631119 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
172538 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56788 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
42412 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
179243 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P152509 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957165 |
AK006607.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957165 |
AK159293.1 |
mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20024058 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023995 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023996 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20024060 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023989 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00547598 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023992 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00210958 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023993 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20024059 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20024056 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20023999 |
- |
- |