Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01235812_m1
See other GM6432 GE Assays ›
Gene Symbol:
Gm6432
Gene Name:
predicted gene 6432
Gene Aliases:
EG623459
Chromosome Location:
Chr.9: 99229376 - 99237239 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01235812_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Gm6432
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001244762.1
NP_001231691.1
4 - 5
544
120
GenBank mRNA
BU939370.1
-
4 - 5
545
119
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0900002917.mm.1
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Recommended Assay
Mus musculus
1700
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2
A_55_P2065815
-
-
SurePrint G3 Mouse Gene Expression Microarray G4852A
A_55_P2065815
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0900024034.mm.1
NM_001244762.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0900024034.mm.1
NM_001244762.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0900024034.mm.1
TC0900002917.mm.1
MTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
558733
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0