Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235831_m1
          See other GRM2 GE Assays ›
          Gene Symbol:
          Grm2
          Gene Name:
          glutamate receptor, metabotropic 2
          Gene Aliases:
          4930441L02Rik, Gprc1b, mGluR2, mGluR7
          Chromosome Location:
          Chr.9: 106644534 - 106656109 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235831_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160353.1 NP_001153825.1 2 - 3 671 82
          GenBank mRNA AY255603.1 - 1 - 2 61 82
          BC115866.1 - 2 - 3 644 40081345 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003015.mm.1

          Set Membership:

          Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
          G-protein coupled glutamate receptor signaling pathway
          chemical synaptic transmission
          glutamate secretion
          regulation of synaptic transmission, glutamatergic
          group II metabotropic glutamate receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          calcium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171348 BC115866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171348 BQ174642.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171348 AK046803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2171348 NM_001160353.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871949 AK038373.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11251 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151823 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133577 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171348 BC115866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171348 BQ174642.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171348 AK046803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2171348 NM_001160353.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133577 - -
          Whole Mouse Genome G4122A A_51_P414219 AK015353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494725 AK034263.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151823 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08131738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07973800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07973809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07853648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07973810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26804306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07503342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08131748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596532 - -
          Affymetrix HT MG-430 PM Array Plate 1431344_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435607_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596521 - -
          Mouse Genome 430 2.0 Array 1431344_PM_at - -
          Mouse Genome 430 2.0 Array 1435607_at - -
          Mouse Genome 430 2.0 Array 1435607_PM_at - -
          Mouse Genome 430 2.0 Array 1431344_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108068_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108068_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900025078.mm.1 NM_001160353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025078.mm.1 NM_001160353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025078.mm.1 TC0900003015.mm.1 MTA TC
          Mouse Expression Set 430 1431344_PM_at - -
          Mouse Expression Set 430 1431344_at - -
          Mouse Expression Set 430 1435607_at - -
          Mouse Expression Set 430 1435607_PM_at - -
          Murine Genome U74v2 Set 171603_at - -
          Murine Genome U74v2 Set 171603_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435607_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435607_at - -
          Affymetrix HT Mouse Genome 430B Array 1431344_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431344_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105220551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline