Search
Search

Iqcf3| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026645.3 | NP_080921.2 | 6 - 7 | 601 | 73 | |
| GenBank mRNA | AK016688.1 | - | 6 - 7 | 602 | 73 | |
| AK018835.1 | - | 6 - 7 | 604 | 73 | ||
| AK132739.1 | - | 6 - 7 | 606 | 73 | ||
| BC049549.1 | - | 6 - 7 | 602 | 6774389 | 73 | |
| DQ153247.1 | - | 6 - 7 | 607 | 73 | ||
Set Membership: |
|