Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236543_g1
          See other TBX10 GE Assays ›
          Gene Symbol:
          Tbx10
          Gene Name:
          T-box 10
          Gene Aliases:
          Dc, Tbx13, Tbx7
          Chromosome Location:
          Chr.19: 3992752 - 3999512 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236543_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbx10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001320.1 NP_001001320.1 4 - 5 902 65
          NM_011533.2 NP_035663.1 4 - 5 745 65
          GenBank mRNA AB221655.1 - 3 - 4 546 65
          AY229976.1 - 4 - 5 902 65
          AY542280.1 - 4 - 5 745 65
          BC109150.2 - 4 - 5 804 40046608 65
          BC109151.2 - 4 - 5 804 40046618 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000023.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbx10

          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900000253.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000253.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000253.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000254.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000254.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000254.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000254.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000254.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000252.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000252.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000252.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000252.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000252.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000256.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000256.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000256.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000256.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000256.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000257.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000257.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000257.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000257.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000257.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000251.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000251.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000251.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000251.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000251.mm.1 TC1900000023.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000258.mm.1 NM_001001320.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000258.mm.1 NM_001001320.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000258.mm.1 NM_011533.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000258.mm.1 NM_011533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000258.mm.1 TC1900000023.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 783261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0