Search
Search
| Gene Symbol: Ehd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010119.5 | NP_034249.1 | 2 - 3 | 581 | 70 | |
| GenBank mRNA | AF099186.1 | - | 2 - 3 | 584 | 70 | |
| AF173156.1 | - | 2 - 3 | 592 | 70 | ||
| AK012896.1 | - | 2 - 3 | 557 | 70 | ||
| AK077536.1 | - | 2 - 3 | 561 | 70 | ||
| AK088980.1 | - | 3 - 4 | 723 | 70 | ||
| AK152821.1 | - | 2 - 3 | 530 | 70 | ||
| AK154651.1 | - | 2 - 3 | 570 | 70 | ||
| AK159191.1 | - | 2 - 3 | 556 | 70 | ||
| AK159943.1 | - | 2 - 3 | 566 | 70 | ||
| AK160535.1 | - | 2 - 3 | 569 | 70 | ||
| AK166701.1 | - | 2 - 3 | 567 | 70 | ||
| AK167721.1 | - | 2 - 3 | 573 | 70 | ||
| AK168610.1 | - | 2 - 3 | 557 | 70 | ||
| AK170679.1 | - | 2 - 3 | 357 | 70 | ||
| AK171047.1 | - | 2 - 3 | 585 | 70 | ||
| AK172539.1 | - | 2 - 3 | 557 | 70 | ||
| AK172669.1 | - | 2 - 3 | 743 | 70 | ||
| BC037094.1 | - | 2 - 3 | 568 | 70 | ||
| BC043332.1 | - | 2 - 3 | 551 | 4981832 | 70 | |
| BC054828.1 | - | 2 - 3 | 545 | 5042951 | 70 | |
Set Membership: |
|