Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236878_m1
          See other ACSBG2 GE Assays ›
          Gene Symbol:
          Acsbg2
          Gene Name:
          acyl-CoA synthetase bubblegum family member 2
          Gene Aliases:
          Bgr
          Chromosome Location:
          Chr.17: 56843103 - 56874504 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236878_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsbg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039114.1 NP_001034203.1 2 - 3 350 61
          GenBank mRNA DQ250679.1 - 2 - 3 350 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002338.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          fatty acid beta-oxidation
          multicellular organism development
          spermatogenesis
          metabolic process
          cell differentiation
          nucleotide binding
          catalytic activity
          long-chain fatty acid-CoA ligase activity
          ATP binding
          ligase activity
          acyl-CoA hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993723 NM_001039114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993723 DQ250679.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51340 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993723 NM_001039114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993723 DQ250679.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31258866 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812658 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03340380 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812642 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812612 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08812624 - -
          Whole Mouse Genome G4122A A_52_P357901 NM_001039114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357901 DQ250679.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08812612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31258784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13226388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13473229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13056911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13276879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13473228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13514820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13156283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31258814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10452175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452176 - -
          Affymetrix HT MG-430 PM Array Plate 1439000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10452172 - -
          Mouse Genome 430 2.0 Array 1439000_at - -
          Mouse Genome 430 2.0 Array 1439000_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328845_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328845_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700021913.mm.1 NM_001039114.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021913.mm.1 NM_001039114.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021913.mm.1 TC1700002338.mm.1 MTA TC
          Mouse Expression Set 430 1439000_PM_at - -
          Mouse Expression Set 430 1439000_at - -
          Murine Genome U74v2 Set 166378_at - -
          Murine Genome U74v2 Set 166378_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439000_at - -
          Affymetrix HT Mouse Genome 430B Array 1439000_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382206 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250547 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2842155 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2842155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline