Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017393.2 | NP_059089.1 | 1 - 2 | 251 | 74 | |
GenBank mRNA | AJ005253.1 | - | 1 - 2 | 219 | 74 | |
AK002238.1 | - | 1 - 2 | 190 | 74 | ||
AK004024.1 | - | 1 - 2 | 200 | 74 | ||
AK145765.1 | - | 1 - 2 | 213 | 74 | ||
AK147719.1 | - | 1 - 2 | 179 | 74 | ||
AK168053.1 | - | 1 - 2 | 198 | 74 | ||
BC001998.1 | - | 1 - 2 | 210 | 3487489 | 74 | |
BY294002.1 | - | 1 - 2 | 252 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |