Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236979_g1
          See other EIF2S3X GE Assays ›
          Gene Symbol:
          Eif2s3x
          Gene Name:
          eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked
          Gene Aliases:
          AA409828, AA547477, AI314668, Eif-2gx
          Chromosome Location:
          Chr.X: 94188709 - 94212651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236979_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif2s3x
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012010.3 NP_036140.1 6 - 7 654 148
          GenBank mRNA AJ006587.1 - 6 - 7 644 148
          AK010819.1 - 3 - 4 332 148
          AK128949.1 - 6 - 7 653 148
          AK146900.1 - 6 - 7 695 148
          AK150095.1 - 6 - 7 661 148
          AK151579.1 - 2 - 3 210 148
          AK153568.1 - 2 - 3 210 148
          AK165866.1 - 6 - 7 656 148
          AK171317.1 - 6 - 7 655 148
          AK208120.1 - 2 - 3 148
          BC036993.1 - 6 - 7 652 148
          BC063755.1 - 6 - 7 654 6513251 148
          BC139234.1 - 6 - 7 654 8862256 148
          BC139235.1 - 6 - 7 654 8862257 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif2s3x

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational initiation
          nucleotide binding
          translation initiation factor activity
          GTPase activity
          GTP binding
          translation factor activity, RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00016528.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016528.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016528.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016533.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016533.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016533.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016537.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016537.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016537.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016535.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016535.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016535.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016538.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016538.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016538.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016530.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016530.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016530.mm.1 TC0X00002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016531.mm.1 NM_012010.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016531.mm.1 NM_012010.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016531.mm.1 TC0X00002643.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline