Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01237023_m1
          See other AMDHD1 GE Assays ›
          Gene Symbol:
          Amdhd1
          Gene Name:
          amidohydrolase domain containing 1
          Gene Aliases:
          1300019J08Rik
          Chromosome Location:
          Chr.10: 93523338 - 93540043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01237023_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amdhd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027908.1 NP_082184.1 5 - 6 878 61
          GenBank mRNA AK005066.1 - 5 - 6 834 61
          AK090143.1 - 5 - 6 869 61
          BC022626.1 - 5 - 6 756 61
          BC043681.1 - 5 - 6 811 61
          BC055025.1 - 5 - 6 776 61
          BC118616.1 - 5 - 6 865 40107296 61
          BC121826.1 - 5 - 6 864 40107295 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002695.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          metalloprotease protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          histidine metabolic process
          histidine catabolic process to glutamate and formamide
          hydrolase activity
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
          metal ion binding
          imidazolonepropionase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 BC043681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 NM_027908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 AK005066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 AK090143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 BC043681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 NM_027908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 AK005066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196862 AK090143.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475250 AK030099.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475250 AK134479.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475250 BC058081.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475250 BC071192.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475250 NM_172403.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61302 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125673 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126961 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 BC043681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 NM_027908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 AK005066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 AK090143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 BC043681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 NM_027908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 AK005066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196862 AK090143.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125673 - -
          Whole Mouse Genome G4122A A_51_P196862 BC043681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 NM_027908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 AK005066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 AK090143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 BC043681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 NM_027908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 AK005066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196862 AK090143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399204 BC118616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399204 BC121826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399204 BC022626.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399204 BC043681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399204 BC055025.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125673 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126961 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371975 - -
          Affymetrix HT MG-430 PM Array Plate 1427370_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447380_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371970 - -
          Mouse Genome 430 2.0 Array 1427370_at - -
          Mouse Genome 430 2.0 Array 1447380_PM_at - -
          Mouse Genome 430 2.0 Array 1427370_PM_at - -
          Mouse Genome 430 2.0 Array 1447380_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71761_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71761_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000020672.mm.1 NM_027908.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020672.mm.1 NM_027908.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020672.mm.1 TC1000002695.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020671.mm.1 NM_027908.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020671.mm.1 NM_027908.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020671.mm.1 TC1000002695.mm.1 MTA TC
          Mouse Expression Set 430 1427370_at - -
          Mouse Expression Set 430 1427370_PM_at - -
          Mouse Expression Set 430 1447380_PM_at - -
          Mouse Expression Set 430 1447380_at - -
          Mouse Genome 430A 2.0 Array 1427370_PM_at - -
          Mouse Genome 430A 2.0 Array 1427370_at - -
          Affymetrix HT Mouse Genome 430A Array 1427370_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427370_at - -
          Affymetrix HT Mouse Genome 430B Array 1447380_at - -
          Affymetrix HT Mouse Genome 430B Array 1447380_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 525100 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232159 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0