Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238131_s1
          See other MAGEE2 GE Assays ›
          Gene Symbol:
          Magee2
          Gene Name:
          melanoma antigen, family E, 2
          Gene Aliases:
          9630059J11Rik, BB131182, Mage-e2
          Chromosome Location:
          Chr.X: 104854952 - 104857267 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238131_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Magee2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053206.2 NP_444436.1 1 1643 118
          GenBank mRNA AF319982.1 - 1 285 118
          BC094361.1 - 1 1532 6829915 118
          BC132640.1 - 1 1498 40130917 118
          BC138210.1 - 1 1498 8861229 118

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Magee2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578577 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349864 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146356 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139212 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353543 AF319982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32578569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146356 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139212 - -
          Whole Mouse Genome G4122A A_52_P383105 BC132640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P383105 BC138210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P383105 AK036343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P383105 AK134670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P383105 BC094361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353543 AF319982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353543 AF319982.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129027 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139212 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14712470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14712453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14712444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14712449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14609990 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14712460 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32578579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32578537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14668343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14668355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14668352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14609981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14609990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14687151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14609982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14668348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14712452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14712463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14712470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32578604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606251 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606248 - -
          Affymetrix Mouse Clariom S Array PSR0X00017680.mm.1 NM_053206.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017680.mm.1 NM_053206.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017680.mm.1 TC0X00002805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017681.mm.1 NM_053206.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017681.mm.1 NM_053206.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017681.mm.1 TC0X00002805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017683.mm.1 NM_053206.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017683.mm.1 NM_053206.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017683.mm.1 TC0X00002805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017682.mm.1 NM_053206.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017682.mm.1 NM_053206.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017682.mm.1 TC0X00002805.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 480314 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2477690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2477690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline