Search
Search

Tmem151a| Gene Symbol: Tmem151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001885.1 | NP_001001885.1 | 1 - 2 | 183 | 69 | |
| GenBank mRNA | AK139456.1 | - | 1 - 2 | 238 | 69 | |
| AK144531.1 | - | 1 - 2 | 196 | 69 | ||
| AK155690.1 | - | 1 - 2 | 198 | 69 | ||
| AK163651.1 | - | 1 - 2 | 200 | 69 | ||
| BC072634.1 | - | 1 - 2 | 183 | 6850125 | 69 | |
Set Membership: |
|