Search
Search

Dnmt3b| Gene Symbol: Dnmt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003960.4 | NP_001003960.2 | 19 - 20 | 2518 | 83 | |
| NM_001003961.4 | NP_001003961.2 | 20 - 21 | 2578 | 83 | ||
| NM_001003963.4 | NP_001003963.2 | 20 - 21 | 2578 | 83 | ||
| NM_001122997.2 | NP_001116469.1 | 19 - 20 | 2466 | 83 | ||
| NM_001271744.1 | NP_001258673.1 | 19 - 20 | 2463 | 83 | ||
| NM_001271745.1 | NP_001258674.1 | 18 - 19 | 2403 | 83 | ||
| NM_001271746.1 | NP_001258675.1 | 19 - 20 | 2463 | 83 | ||
| NM_001271747.1 | NP_001258676.1 | 18 - 19 | 2403 | 83 | ||
| NM_010068.5 | NP_034198.3 | 19 - 20 | 2518 | 83 | ||
| GenBank mRNA | AF068626.2 | - | 20 - 21 | 2433 | 83 | |
| AF068627.2 | - | 19 - 20 | 2373 | 83 | ||
| AF068628.2 | - | 19 - 20 | 2373 | 83 | ||
| AF151969.1 | - | 20 - 21 | 2576 | 83 | ||
| AF151970.1 | - | 19 - 20 | 2516 | 83 | ||
| AF151971.1 | - | 19 - 20 | 2516 | 83 | ||
| AF151972.1 | - | 20 - 21 | 2576 | 83 | ||
| AF151973.1 | - | 19 - 20 | 2461 | 83 | ||
| AF151974.1 | - | 18 - 19 | 2401 | 83 | ||
| AF151975.1 | - | 19 - 20 | 2461 | 83 | ||
| AF151976.1 | - | 18 - 19 | 2401 | 83 | ||
| AY078427.1 | - | 20 - 21 | 2433 | 83 | ||
| BC105677.1 | - | 19 - 20 | 2298 | 40048574 | 83 | |
| BC105922.1 | - | 19 - 20 | 2298 | 40048584 | 83 | |
Set Membership: |
|