Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240399_m1
          See other SCARF1 GE Assays ›
          Gene Symbol:
          Scarf1
          Gene Name:
          scavenger receptor class F, member 1
          Gene Aliases:
          AA986099, SREC, SREC-I, mKIAA0149
          Chromosome Location:
          Chr.11: 75513495 - 75526580 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240399_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scarf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004157.2 NP_001004157.2 3 - 4 312 70
          GenBank mRNA AK133477.1 - 3 - 4 295 70
          AK220215.1 - 3 - 4 296 70
          BC057600.1 - 2 - 3 219 70
          BC080288.1 - 3 - 4 312 6411836 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001071.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scarf1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          cell adhesion
          positive regulation of neuron projection development
          neuron remodeling
          dendrite development
          positive regulation of axon regeneration
          scavenger receptor activity
          protein binding
          low-density lipoprotein particle binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926564 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958976 AK133477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958976 NM_001004157.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170175 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140826 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958976 AK133477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958976 NM_001004157.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27926546 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07400526 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140826 - -
          Whole Mouse Genome G4122A A_52_P95544 BC080288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95544 BC057600.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95544 BC019441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95544 AK133477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95544 NM_001004157.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140826 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09135115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09135105 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27926564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27926579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08991925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09135116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08991928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08991927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09135133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09135132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09464454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08991930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09135124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09392888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09135125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27926614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378642 - -
          Affymetrix HT MG-430 PM Array Plate 1434490_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378637 - -
          Mouse Genome 430 2.0 Array 1440748_at - -
          Mouse Genome 430 2.0 Array 1440748_PM_at - -
          Mouse Genome 430 2.0 Array 1434490_PM_at - -
          Mouse Genome 430 2.0 Array 1434490_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380713_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380713_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100009711.mm.1 NM_001004157.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009711.mm.1 NM_001004157.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009711.mm.1 TC1100001071.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430B Array 1434490_at - -
          Affymetrix HT Mouse Genome 430B Array 1434490_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 744643 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2534249 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2820703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2820703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline