Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240517_m1
          See other CCDC153 GE Assays ›
          Gene Symbol:
          Ccdc153
          Gene Name:
          coiled-coil domain containing 153
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 44240655 - 44247306 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240517_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc153
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081369.2 NP_001074838.2 5 - 6 401 65
          GenBank mRNA BC038167.1 - 3 - 4 452 65
          CX240611.1 - 5 - 6 378 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000503.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 BC038167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 BG072259.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 CX240611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 NM_001081369.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 BC038167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 BG072259.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 CX240611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P352735 NM_001081369.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95945 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 BC038167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 BG072259.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 CX240611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 NM_001081369.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 BC038167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 BG072259.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 CX240611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P352735 NM_001081369.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01918211 - -
          Whole Mouse Genome G4122A A_52_P352735 BC038167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 BG072259.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 CX240611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 NM_001081369.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 BC038167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 BG072259.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 CX240611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P352735 NM_001081369.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26466524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07952986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07952992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07912720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08036698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08098332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07704384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07912717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07799340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08098335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07704383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07952983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07953514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26466463 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584697 - -
          Affymetrix HT MG-430 PM Array Plate 1458104_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10584691 - -
          Mouse Genome 430 2.0 Array 1458104_PM_a_at - -
          Mouse Genome 430 2.0 Array 1458104_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270150_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270150_at - -
          Affymetrix Mouse Clariom S Array PSR0900003387.mm.1 NM_001081369.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003387.mm.1 NM_001081369.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003387.mm.1 TC0900000503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003390.mm.1 NM_001081369.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003390.mm.1 NM_001081369.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003390.mm.1 TC0900000503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003389.mm.1 NM_001081369.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003389.mm.1 NM_001081369.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003389.mm.1 TC0900000503.mm.1 MTA TC
          Mouse Expression Set 430 1458104_PM_a_at - -
          Mouse Expression Set 430 1458104_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1458104_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1458104_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 657197 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223757 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline