Search
Search

Kcng4| Gene Symbol: Kcng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025734.2 | NP_080010.2 | 2 - 3 | 1234 | 75 | |
| GenBank mRNA | AF450109.1 | - | 2 - 3 | 900 | 75 | |
| BC043936.1 | - | 2 - 3 | 1234 | 4504272 | 75 | |
Set Membership: |
|