Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01241477_m1
          See other DEFB21 GE Assays ›
          Gene Symbol:
          Defb21
          Gene Name:
          defensin beta 21
          Gene Aliases:
          4930525K10Rik, Gm121
          Chromosome Location:
          Chr.2: 152572744 - 152574944 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01241477_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207276.2 NP_997159.1 2 - 3 166 80
          GenBank mRNA AK076875.1 - 2 - 3 168 80
          DQ012032.1 - 2 - 3 166 80
          DQ141308.1 - 1 - 2 87 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002290.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb21

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          defense response to bacterium
          innate immune response
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107399 NM_207276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107399 AK076875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107399 DQ012032.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415334 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114196 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107399 NM_207276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107399 AK076875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107399 DQ012032.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749948 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114196 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107399 NM_207276.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107399 AK076875.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107399 DQ012032.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114196 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01397368 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437336 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01397365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01397367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01427537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01503854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01397367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01681249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01427530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01437336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01149870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01503859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01643263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477132 - -
          Affymetrix HT MG-430 PM Array Plate 1447395_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10477129 - -
          Mouse Genome 430 2.0 Array 1447395_PM_at - -
          Mouse Genome 430 2.0 Array 1447395_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 403172_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 403172_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200017910.mm.1 NM_207276.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017910.mm.1 NM_207276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017910.mm.1 TC0200002290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017911.mm.1 NM_207276.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017911.mm.1 NM_207276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017911.mm.1 TC0200002290.mm.1 MTA TC
          Mouse Expression Set 430 1447395_at - -
          Mouse Expression Set 430 1447395_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447395_at - -
          Affymetrix HT Mouse Genome 430B Array 1447395_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 875478 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3390592 - -
          Illumina mouse-6 v1.0 expression beadchip 3390592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2684681 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2684681 - -
          mouse-6 v1.1 (Illumina) scl0403172.2_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 3390592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0403172.2_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2684681 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2684681 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline