| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10582355 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582353 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582348 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582341 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582357 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582363 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582354 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582344 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582362 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582345 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582356 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582346 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582359 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582342 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582343 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582366 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582350 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582367 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582365 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582352 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582360 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582340 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582361 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582338 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582347 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582364 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582349 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582358 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582339 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10582351 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1455157_PM_a_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10582378 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10582337 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10582376 |
- |
- |
| Mouse Genome 430 2.0 Array |
1439387_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439387_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1428350_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1428350_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455157_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455157_PM_a_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800023769.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023769.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023769.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023775.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023775.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023775.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023763.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023763.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023763.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023780.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023780.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023780.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023749.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023749.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023749.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023779.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023779.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023779.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023758.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023758.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023758.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023764.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023764.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023764.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023771.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023771.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023771.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023784.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023784.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023784.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023740.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023740.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023740.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023762.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023762.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023762.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023755.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023755.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023755.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023761.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023761.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023761.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023767.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023767.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023767.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023806.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023806.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023806.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023770.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023770.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023770.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023805.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023805.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023805.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023807.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023807.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023807.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023766.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023766.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023766.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023777.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023777.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023777.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023786.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023786.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023786.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023781.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023781.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023781.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023733.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023733.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023733.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023735.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023735.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023735.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023772.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023772.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023772.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023773.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023773.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023773.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023778.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023778.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023778.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023751.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023751.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023751.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023741.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023741.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023741.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023736.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023736.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023736.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023734.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023734.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023734.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023798.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023798.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023798.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023790.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023790.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023790.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023801.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023801.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023801.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023797.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023797.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023797.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023800.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023800.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023800.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023782.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023782.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023782.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023776.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023776.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023776.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023743.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023743.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023743.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023788.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023788.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023788.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023803.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023803.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023803.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023765.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023765.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023765.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023799.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023799.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023799.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023760.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023760.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023760.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023794.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023794.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023794.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023738.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023738.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023738.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023804.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023804.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023804.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023746.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023746.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023746.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023796.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023796.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023796.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023789.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023789.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023789.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023748.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023748.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023748.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023754.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023754.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023754.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023768.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023768.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023768.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023792.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023792.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023792.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023787.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023787.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023787.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023759.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023759.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023759.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023785.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023785.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023785.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023791.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023791.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023791.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023793.mm.1 |
NM_001037298.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023793.mm.1 |
NM_001037298.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023793.mm.1 |
TC0800003074.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1455157_a_at |
- |
- |
| Mouse Expression Set 430 |
1428350_at |
- |
- |
| Mouse Expression Set 430 |
1439387_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1439387_x_at |
- |
- |
| Mouse Expression Set 430 |
1428350_PM_at |
- |
- |
| Mouse Expression Set 430 |
1455157_PM_a_at |
- |
- |
| Murine Genome U74v2 Set |
170002_at |
- |
- |
| Murine Genome U74v2 Set |
170002_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455157_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455157_PM_a_at |
- |
- |