Search
Search

Fbxw24| Gene Symbol: Fbxw24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013776.4 | NP_001013798.2 | 7 - 8 | 1016 | 81 | |
| GenBank mRNA | BC080701.1 | - | 7 - 8 | 1017 | 30473126 | 81 |
| CO808277.1 | - | 2 - 3 | 364 | 81 | ||
| DV651634.1 | - | 3 - 4 | 454 | 81 | ||
| GQ418172.1 | - | 7 - 8 | 1017 | 81 | ||
Set Membership: |
|