Search
Search

Rap1gap2| Gene Symbol: Rap1gap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015046.2 | NP_001015046.1 | 12 - 13 | 1075 | 59 | |
| GenBank mRNA | AK038834.1 | - | 2 - 3 | 138 | 59 | |
| AK147290.1 | - | 13 - 14 | 1057 | 59 | ||
| BC107388.2 | - | 3 - 4 | 281 | 40057754 | 59 | |
Set Membership: |
|