Search
Search

Baiap2| Gene Symbol: Baiap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037754.3 | NP_001032843.1 | 6 - 7 | 594 | 66 | |
| NM_001037755.3 | NP_001032844.2 | 6 - 7 | 594 | 66 | ||
| NM_130862.4 | NP_570932.2 | 6 - 7 | 594 | 66 | ||
| GenBank mRNA | AF390178.1 | - | 6 - 7 | 552 | 66 | |
| AF390179.1 | - | 6 - 7 | 552 | 66 | ||
| AK049469.1 | - | 6 - 7 | 595 | 66 | ||
| AK145924.1 | - | 6 - 7 | 595 | 66 | ||
| AK158579.1 | - | 6 - 7 | 619 | 66 | ||
| AK160401.1 | - | 6 - 7 | 595 | 66 | ||
| BC006620.1 | - | 6 - 7 | 585 | 3158004 | 66 | |
| BC016411.1 | - | 6 - 7 | 585 | 3158004 | 66 | |
Set Membership: |
|