Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242213_m1
          See other RNF166 GE Assays ›
          Gene Symbol:
          Rnf166
          Gene Name:
          ring finger protein 166
          Gene Aliases:
          1110031E24Rik, AW555115, Zfp313l
          Chromosome Location:
          Chr.8: 122466145 - 122476064 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf166
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033142.2 NP_001028314.1 5 - 6 100
          GenBank mRNA AK151813.1 - 5 - 6 100
          BC131924.1 - 5 - 6 695 40130201 100
          BC132281.1 - 5 - 6 695 40130558 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003072.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK216687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK079883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK151813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 NM_001033142.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK216687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK079883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 AK151813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342535 NM_001033142.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35946 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK216687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK079883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK151813.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 NM_001033142.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK216687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK079883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 AK151813.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342535 NM_001033142.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233614 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06669579 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01848716 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06669591 - -
          Whole Mouse Genome G4122A A_51_P342535 AK216687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 AK079883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 AK151813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 NM_001033142.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 AK216687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 AK079883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 AK151813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342535 NM_001033142.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01848716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07180083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06926007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06926016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06926008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06764095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06836919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06764866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06726515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07209311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06764871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582334 - -
          Affymetrix HT MG-430 PM Array Plate 1436906_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448079_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582330 - -
          Mouse Genome 430 2.0 Array 1448079_PM_at - -
          Mouse Genome 430 2.0 Array 1436906_at - -
          Mouse Genome 430 2.0 Array 1448079_at - -
          Mouse Genome 430 2.0 Array 1457238_PM_at - -
          Mouse Genome 430 2.0 Array 1436906_PM_at - -
          Mouse Genome 430 2.0 Array 1457238_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68718_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68718_at - -
          Affymetrix Mouse Clariom S Array PSR0800023717.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023717.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023717.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023718.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023718.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023718.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023726.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023726.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023726.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023725.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023725.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023725.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023720.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023720.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023720.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023721.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023721.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023721.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023724.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023724.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023724.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023719.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023719.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023719.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023716.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023716.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023716.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023714.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023714.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023714.mm.1 TC0800003072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800023728.mm.1 NM_001033142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023728.mm.1 NM_001033142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800023728.mm.1 TC0800003072.mm.1 MTA TC
          Mouse Expression Set 430 1436906_PM_at - -
          Mouse Expression Set 430 1436906_at - -
          Mouse Expression Set 430 1448079_PM_at - -
          Mouse Expression Set 430 1457238_at - -
          Mouse Expression Set 430 1448079_at - -
          Mouse Expression Set 430 1457238_PM_at - -
          Murine Genome U74v2 Set 98057_PM_at - -
          Murine Genome U74v2 Set 98057_at - -
          Mouse Genome 430A 2.0 Array 1436906_at - -
          Mouse Genome 430A 2.0 Array 1436906_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436906_at - -
          Affymetrix HT Mouse Genome 430A Array 1436906_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1448079_at - -
          Affymetrix HT Mouse Genome 430B Array 1448079_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 458970 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5570333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217188 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217188 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0