Search
Search

Zzef1| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045536.2 | NP_001039001.1 | 34 - 35 | 5500 | 53 | |
| GenBank mRNA | AK134006.1 | - | 13 - 14 | 2062 | 53 | |
Set Membership: |
|