Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242666_m1
          See other 1810043H04RIK GE Assays ›
          Gene Symbol:
          1810043H04Rik
          Gene Name:
          RIKEN cDNA 1810043H04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 120098934 - 120100424 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242666_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810043H04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110242.1 NP_001103712.1 2 - 3 246 54
          GenBank mRNA AK007756.1 - 2 - 3 244 54
          AK010111.1 - 2 - 3 244 54
          BC147711.1 - 2 - 3 226 9087470 54
          BC147720.1 - 2 - 3 226 9087479 54
          BY079397.1 - 2 - 3 253 54

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100050 BC147711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100050 BC147720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100050 AK007756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100050 NM_001110242.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271503 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100050 BC147711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100050 BC147720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100050 AK007756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100050 NM_001110242.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07515400 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100050 BC147711.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100050 BC147720.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100050 AK007756.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100050 NM_001110242.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28182359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28182354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09443412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09465994 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457826_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1448003_PM_at - -
          Mouse Genome 430 2.0 Array 1457826_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448003_PM_at - -
          Mouse Genome 430 2.0 Array 1448003_at - -
          Mouse Genome 430 2.0 Array 1457826_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208501_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208501_at - -
          Affymetrix Mouse Clariom S Array PSR1100019378.mm.1 TC1100002018.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019377.mm.1 NM_001110242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019377.mm.1 NM_001110242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019377.mm.1 TC1100002018.mm.1 MTA TC
          Mouse Expression Set 430 1457826_a_at - -
          Mouse Expression Set 430 1448003_PM_at - -
          Mouse Expression Set 430 1457826_PM_a_at - -
          Mouse Expression Set 430 1448003_at - -
          Affymetrix HT Mouse Genome 430B Array 1457826_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1457826_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1448003_at - -
          Affymetrix HT Mouse Genome 430B Array 1448003_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102970603 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245809 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline