Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242886_g1
          See other PMCH GE Assays ›
          Gene Symbol:
          Pmch
          Gene Name:
          pro-melanin-concentrating hormone
          Gene Aliases:
          A230109K23Rik
          Chromosome Location:
          Chr.10: 88091072 - 88092374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242886_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pmch
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029971.2 NP_084247.1 1 - 2 310 78
          GenBank mRNA AK020723.1 - 1 - 2 310 78
          AK138363.1 - 1 - 2 313 78
          BC048534.1 - 1 - 2 310 6533154 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001072.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pmch

          Panther Classification:

          Molecular Function -

          peptide hormone intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          chemical synaptic transmission
          feeding behavior
          response to cold
          negative regulation of synaptic transmission, dopaminergic
          glucose homeostasis
          drinking behavior
          negative regulation of blood pressure
          positive regulation of circadian sleep/wake cycle, REM sleep
          regulation of neuronal synaptic plasticity
          receptor binding
          hormone activity
          melanin-concentrating hormone activity
          type 1 melanin-concentrating hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 NM_029971.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 AK020723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 AK138363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 BB169239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 NM_029971.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 AK020723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 AK138363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 BB169239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125566 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138116 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 NM_029971.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 AK020723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 AK138363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 BB169239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 BC048534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 NM_029971.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 AK020723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 AK138363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 BB169239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 NM_029971.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 AK020723.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 AK138363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 BB169239.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 NM_029971.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 AK020723.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 AK138363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 BB169239.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27323529 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125566 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138116 - -
          Whole Mouse Genome G4122A A_52_P418814 NM_029971.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 AK020723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 AK138363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 BB169239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 BC048534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 NM_029971.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 AK020723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 AK138363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 BB169239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P418814 BC048534.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125566 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138116 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08456661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08456664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08456672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27323472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08845576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27323512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365576 - -
          Affymetrix HT MG-430 PM Array Plate 1429361_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371591 - -
          Affymetrix Mouse Gene 1.1 ST Array 10365574 - -
          Mouse Genome 430 2.0 Array 1429361_PM_at - -
          Mouse Genome 430 2.0 Array 1429361_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110312_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110312_at - -
          Affymetrix Mouse Clariom S Array PSR1000008099.mm.1 NM_029971.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008099.mm.1 NM_029971.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008099.mm.1 TC1000001072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000008100.mm.1 NM_029971.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008100.mm.1 NM_029971.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008100.mm.1 TC1000001072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000008098.mm.1 NM_029971.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008098.mm.1 NM_029971.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000008098.mm.1 TC1000001072.mm.1 MTA TC
          Mouse Expression Set 430 1429361_PM_at - -
          Mouse Expression Set 430 1429361_at - -
          Affymetrix HT Mouse Genome 430B Array 1429361_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429361_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 535238 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2421388 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2955329 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2955329 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline