Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243354_m1
          See other NUP85 GE Assays ›
          Gene Symbol:
          Nup85
          Gene Name:
          nucleoporin 85
          Gene Aliases:
          Pcnt1, frount
          Chromosome Location:
          Chr.11: 115564432 - 115583924 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002929.4 NP_001002929.3 14 - 15 1483 59
          GenBank mRNA AF498263.1 - 14 - 15 1397 59
          AK017632.1 - 14 - 15 1448 59
          BC079856.1 - 14 - 15 1458 6403446 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001899.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nup85

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          chemotaxis
          protein transport
          cytokine-mediated signaling pathway
          lamellipodium assembly
          macrophage chemotaxis
          mRNA transport
          CCR2 chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981296 AK017632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981296 BC079856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981296 CF738553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981296 NM_001002929.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47463 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107100 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104261 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981296 AK017632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981296 BC079856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981296 CF738553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981296 NM_001002929.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201532 AK017632.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201532 BC079856.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201532 CF738553.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201532 NM_001002929.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28154148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104261 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07502865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07502850 - -
          Whole Mouse Genome G4122A A_51_P201532 AK017632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P201532 BC079856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P201532 CF738553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P201532 NM_001002929.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127460 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107100 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09319003 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09318991 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09319001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09318995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28154116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09530035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09319005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09678780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09200266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09296569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09318992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09296571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09448935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09200264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09296570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09530027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28154155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382547 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382544 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382558 - -
          Affymetrix HT MG-430 PM Array Plate 1416073_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1437520_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382542 - -
          Mouse Genome 430 2.0 Array 1437520_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416073_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437520_a_at - -
          Mouse Genome 430 2.0 Array 1416073_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 445007_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 445007_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100017892.mm.1 NM_001002929.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017892.mm.1 NM_001002929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017892.mm.1 TC1100001899.mm.1 MTA TC
          Mouse Expression Set 430 1437520_a_at - -
          Mouse Expression Set 430 1437520_PM_a_at - -
          Murine Genome U74v2 Set 99662_at - -
          Murine Genome U74v2 Set 99663_PM_g_at - -
          Murine Genome U74v2 Set 99663_g_at - -
          Murine Genome U74v2 Set 99662_PM_at - -
          Mouse Genome 430A 2.0 Array 1437520_a_at - -
          Mouse Genome 430A 2.0 Array 1437520_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416073_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437520_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416073_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437520_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 722633 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105570044 - -
          Illumina mouse-6 v1.0 expression beadchip 105570044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214923 - -
          Illumina Sentrix mouse-6.v1 bead array 105570044 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105570044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2890574 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2890574 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline