Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243673_m1
          See other INCA1 GE Assays ›
          Gene Symbol:
          Inca1
          Gene Name:
          inhibitor of CDK, cyclin A1 interacting protein 1
          Gene Aliases:
          AI842396
          Chromosome Location:
          Chr.11: 70688361 - 70700206 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243673_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inca1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252482.1 NP_001239411.1 5 - 6 640 75
          NM_001252483.1 NP_001239412.1 5 - 6 601 75
          NM_001252484.1 NP_001239413.1 5 - 6 531 75
          NM_001252485.1 NP_001239414.1 6 - 7 760 75
          NM_213729.1 NP_998894.1 5 - 6 669 75
          GenBank mRNA AK139002.1 - 5 - 6 529 75
          AY601909.1 - 5 - 6 669 75
          AY601910.1 - 5 - 6 640 75
          AY601911.1 - 5 - 6 601 75
          AY601912.1 - 5 - 6 531 75
          BC096427.1 - 6 - 7 714 4160694 75
          BC115572.1 - 5 - 6 500 40102153 75
          BC115573.1 - 5 - 6 500 40102154 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Inca1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell proliferation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of apoptotic signaling pathway
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          cyclin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252482.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252482.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252484.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252484.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252485.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_001252485.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_213729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 NM_213729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028761.mm.1 TC1100003111.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline