Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244544_m1
          See other LNCBATE1 GE Assays ›
          Gene Symbol:
          Lncbate1
          Gene Name:
          brown adipose tissue enriched long non-coding RNA 1
          Gene Aliases:
          3010033K07Rik, Lnc-BATE1
          Chromosome Location:
          Chr.8: 108553252 - 108584677 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244544_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lncbate1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_077224.1 1 - 2 523 112
          GenBank mRNA AK080070.1 - 1 - 2 442 112
          DV056598.1 - 1 - 2 542 112
          DV056599.1 - 1 - 2 542 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002887.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63065 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06634709 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01823287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06634686 - -
          Whole Mouse Genome G4122A A_52_P177352 AI181005.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177352 AK080070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P177352 BB219789.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06634686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07258384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06999750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07330792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07092355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06936107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07258377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07027166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07258379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06999756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06685499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07033116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06934488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07150449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07028997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06964808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07084266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07398912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06893350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06903125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26151763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800022238.mm.1 NR_077224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022238.mm.1 TC0800002887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022242.mm.1 NR_077224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022242.mm.1 TC0800002887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022235.mm.1 NR_077224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022235.mm.1 TC0800002887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022239.mm.1 NR_077224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022239.mm.1 TC0800002887.mm.1 MTA TC
          Murine Genome U74v2 Set 131869_at - -
          Murine Genome U74v2 Set 131869_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 689136 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6650136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline