Search
Search

Gm20735| Gene Symbol: Gm20735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015497.2 | 1 - 2 | 154 | 67 | ||
| GenBank mRNA | AK042436.1 | - | 1 - 2 | 154 | 67 | |
| BX522380.1 | - | 1 - 2 | 120 | 67 | ||
Set Membership: |
|