Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244826_g1
          See other CCL17 GE Assays ›
          Gene Symbol:
          Ccl17
          Gene Name:
          chemokine (C-C motif) ligand 17
          Gene Aliases:
          Abcd-2, Scya17, Scya17l, Tarc
          Chromosome Location:
          Chr.8: 94807404 - 94812038 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244826_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011332.3 NP_035462.2 2 - 3 237 65
          GenBank mRNA AF125570.1 - 2 - 3 326 65
          AF125571.1 - 2 - 3 268 65
          AF125572.1 - 2 - 3 241 65
          AJ242587.1 - 2 - 3 216 65
          AK007663.1 - 2 - 3 238 65
          BC028505.1 - 2 - 3 221 3328278 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001121.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl17

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          cellular defense response
          signal transduction
          negative regulation of myoblast differentiation
          cell chemotaxis
          cytokine activity
          chemokine activity
          CCR4 chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71222 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157627 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114462 AJ242587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079563 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157627 - -
          Whole Mouse Genome G4122A A_51_P114462 AJ242587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114462 AJ242587.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133633 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896998 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896989 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06896987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07349432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07018277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06805673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06896979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06805676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06896998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07444471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06805672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07018278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07178717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07078328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06860791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07319242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07319252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06805674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06811487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06896987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06897149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06811484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07091947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574227 - -
          Affymetrix HT MG-430 PM Array Plate 1419413_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574226 - -
          Mouse Genome 430 2.0 Array 1419413_PM_at - -
          Mouse Genome 430 2.0 Array 1419413_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20295_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20295_at - -
          Affymetrix Mouse Clariom S Array PSR0800008739.mm.1 NM_011332.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008739.mm.1 NM_011332.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008739.mm.1 TC0800001121.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008741.mm.1 NM_011332.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008741.mm.1 NM_011332.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008741.mm.1 TC0800001121.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008740.mm.1 NM_011332.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008740.mm.1 NM_011332.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008740.mm.1 TC0800001121.mm.1 MTA TC
          Mouse Expression Set 430 1419413_at - -
          Mouse Expression Set 430 1419413_PM_at - -
          Murine Genome U74v2 Set 97783_at - -
          Murine Genome U74v2 Set 97783_PM_at - -
          Mouse Genome 430A 2.0 Array 1419413_PM_at - -
          Mouse Genome 430A 2.0 Array 1419413_at - -
          Affymetrix HT Mouse Genome 430A Array 1419413_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913884 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 630121 - -
          Illumina mouse-6 v1.0 expression beadchip 630121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717015 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717015 - -
          mouse-6 v1.1 (Illumina) scl33474.3.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 630121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33474.3.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717015 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33474.3.1_12-S - -

          Back To Top
           

          Citations (1)

          Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
          Authors
          Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
          Journal
          J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
          Volume (Year)
          184 (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline