Search
Search

Clasp2| Gene Symbol: Clasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081960.1 | NP_001075429.1 | 2 - 3 | 331 | 135 | |
| NM_001114347.1 | NP_001107819.1 | 8 - 9 | 1073 | 135 | ||
| NM_001286599.1 | NP_001273528.1 | 8 - 9 | 1073 | 135 | ||
| NM_001286600.1 | NP_001273529.1 | 2 - 3 | 331 | 135 | ||
| NM_001286601.1 | NP_001273530.1 | 2 - 3 | 331 | 152 | ||
| NM_001286602.1 | NP_001273531.1 | 2 - 3 | 331 | 135 | ||
| NM_001286603.1 | NP_001273532.1 | 2 - 3 | 287 | 135 | ||
| NM_029633.2 | NP_083909.2 | 2 - 3 | 331 | 135 | ||
| GenBank mRNA | AJ276961.1 | - | 8 - 9 | 1073 | 135 | |
| AK005146.1 | - | 2 - 3 | 283 | 135 | ||
| AK034560.1 | - | 8 - 9 | 1036 | 135 | ||
| AK043551.1 | - | 2 - 3 | 331 | 135 | ||
| AK148966.1 | - | 2 - 3 | 332 | 135 | ||
| AK164326.1 | - | 2 - 3 | 331 | 135 | ||
| BC030468.1 | - | 2 - 3 | 248 | 135 | ||
| BC117963.1 | - | 2 - 3 | 319 | 40098296 | 135 | |
| BC117964.1 | - | 2 - 3 | 319 | 40098298 | 135 | |
| BC131791.1 | - | 2 - 3 | 319 | 40098299 | 135 | |
| BC131792.1 | - | 2 - 3 | 319 | 40098301 | 152 | |
| BC141415.1 | - | 2 - 3 | 317 | 9056028 | 135 | |
| CJ052159.1 | - | 2 - 3 | 288 | 135 | ||
Set Membership: |
|