Search
Search

Trank1| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164659.1 | NP_001158131.1 | 9 - 10 | 1697 | 59 | |
| GenBank mRNA | AK079552.1 | - | 9 - 10 | 1698 | 59 | |
Set Membership: |
|