Search
Search

Sdcbp2| Gene Symbol: Sdcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145535.2 | NP_663510.1 | 7 - 8 | 828 | 67 | |
| GenBank mRNA | AK136572.1 | - | 7 - 8 | 784 | 67 | |
| AK144452.1 | - | 7 - 8 | 793 | 67 | ||
| AK160483.1 | - | 7 - 8 | 786 | 67 | ||
| BC005556.1 | - | 7 - 8 | 819 | 3964815 | 67 | |
Set Membership: |
|