Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246403_g1
          See other TK1 GE Assays ›
          Gene Symbol:
          Tk1
          Gene Name:
          thymidine kinase 1
          Gene Aliases:
          D530002A18Rik, Tk-1, Tk1a, Tk1b
          Chromosome Location:
          Chr.11: 117815509 - 117826014 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246403_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271729.1 NP_001258658.1 4 - 5 599 91
          NM_009387.2 NP_033413.2 5 - 6 631 91
          GenBank mRNA AK085188.1 - 5 - 6 480 91
          AK087988.1 - 5 - 6 434 91
          AK139244.1 - 5 - 6 451 91
          BC012215.1 - 5 - 6 435 3969287 91
          BC091758.1 - 5 - 6 464 4978985 91
          CF732600.1 - 4 - 5 414 91
          M11945.1 - 5 - 6 428 91
          M19438.1 - 4 - 5 321 91
          S57244.1 - 5 - 6 428 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tk1

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          deoxyribonucleoside monophosphate biosynthetic process
          nucleotide biosynthetic process
          phosphorylation
          thymidine metabolic process
          protein homotetramerization
          DNA biosynthetic process
          nucleotide binding
          thymidine kinase activity
          ATP binding
          zinc ion binding
          kinase activity
          transferase activity
          nucleoside kinase activity
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056496 AK139244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056496 BC012215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056496 AK087988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056496 BC091758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056496 NM_009387.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316792 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178656 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139192 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056496 AK139244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056496 BC012215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056496 AK087988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056496 BC091758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056496 NM_009387.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P224013 M19438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P224013 AK139244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P224013 BC012215.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P224013 AK087988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P224013 BC091758.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28167452 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07509086 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156611 - -
          Whole Mouse Genome G4122A A_51_P224013 M19438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224013 AK139244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224013 BC012215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224013 AK087988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224013 BC091758.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09413611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09498498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09498486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28167490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09287291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09287289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09386671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09369341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09432867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09711549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09226717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09137274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09386672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28167461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582810 - -
          Affymetrix HT MG-430 PM Array Plate 1416258_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393431 - -
          Affymetrix Mouse Gene 1.1 ST Array 10582809 - -
          Affymetrix Mouse Gene 1.1 ST Array 10382985 - -
          Mouse Genome 430 2.0 Array 1416258_at - -
          Mouse Genome 430 2.0 Array 1416258_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21877_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21877_at - -
          Affymetrix Mouse Clariom S Array PSR1100039297.mm.1 NM_001271729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039297.mm.1 NM_009387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039297.mm.1 NM_009387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039297.mm.1 TC1100004147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039295.mm.1 NM_001271729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039295.mm.1 NM_009387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039295.mm.1 NM_009387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039295.mm.1 TC1100004147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039296.mm.1 NM_001271729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039296.mm.1 NM_009387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039296.mm.1 NM_009387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039296.mm.1 TC1100004147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039298.mm.1 NM_001271729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039298.mm.1 NM_009387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039298.mm.1 NM_009387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039298.mm.1 TC1100004147.mm.1 MTA TC
          Mouse Expression Set 430 1416258_at - -
          Mouse Expression Set 430 1416258_PM_at - -
          Murine Genome U74v2 Set 96081_at - -
          Murine Genome U74v2 Set 161150_PM_at - -
          Murine Genome U74v2 Set 161150_at - -
          Murine Genome U74v2 Set 96081_PM_at - -
          Murine Genome U74v2 Set 139602_at - -
          Murine Genome U74v2 Set 171407_i_at - -
          Murine Genome U74v2 Set 165988_r_at - -
          Murine Genome U74v2 Set 171407_PM_i_at - -
          Murine Genome U74v2 Set 139602_PM_at - -
          Murine Genome U74v2 Set 165988_PM_r_at - -
          Murine Genome U74v2 Set 171561_at - -
          Murine Genome U74v2 Set 171561_PM_at - -
          Mouse Genome 430A 2.0 Array 1416258_at - -
          Mouse Genome 430A 2.0 Array 1416258_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416258_at - -
          Affymetrix HT Mouse Genome 430A Array 1416258_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 831955 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760152 - -
          Illumina mouse-6 v1.0 expression beadchip 6760152 - -
          Illumina mouse-6 v1.0 expression beadchip 540014 - -
          Illumina mouse-6 v1.0 expression beadchip 540494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611858 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769718 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605890 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769718 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611858 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605890 - -
          mouse-6 v1.1 (Illumina) scl0021877.1_66-S - -
          mouse-6 v1.1 (Illumina) scl021877.4_33-S - -
          mouse-6 v1.1 (Illumina) scl0001554.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 540494 - -
          Illumina Sentrix mouse-6.v1 bead array 540014 - -
          Illumina Sentrix mouse-6.v1 bead array 6760152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0021877.1_66-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl021877.4_33-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001554.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540014 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540494 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611857 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2893926 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605890 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605890 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611857 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611858 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2893926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline