Search
Search

Sgsm2| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197943.2 | NP_922934.2 | 12 - 13 | 1639 | 66 | |
| GenBank mRNA | AK078057.1 | - | 1 - 2 | 829 | 66 | |
| AK122273.1 | - | 12 - 13 | 1561 | 66 | ||
| BC058414.1 | - | 12 - 13 | 1630 | 6822416 | 66 | |
| BC060163.1 | - | 12 - 13 | 1630 | 6822416 | 66 | |
Set Membership: |
|