Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246976_m1
          See other 1700012B07RIK GE Assays ›
          Gene Symbol:
          1700012B07Rik
          Gene Name:
          RIKEN cDNA 1700012B07 gene
          Gene Aliases:
          1700069D21Rik
          Chromosome Location:
          Chr.11: 109788268 - 109828079 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246976_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700012B07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162428.1 NP_001155900.1 4 - 5 702 85
          NM_027038.1 NP_081314.1 4 - 5 702 85
          GenBank mRNA AK005888.1 - 4 - 5 696 85
          AK132924.1 - 703 85
          AK160153.1 - 4 - 5 703 85
          BC048675.1 - 4 - 5 678 6774204 85
          BC099559.1 - 4 - 5 681 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003995.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 BC099559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 BC048675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK005888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK132924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK160153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 BC099559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 BC048675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK005888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK132924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P287241 AK160153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983688 AK132924.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143311 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180223 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118499 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 BC099559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 BC048675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK005888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK132924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK160153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 BC099559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 BC048675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK005888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK132924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P287241 AK160153.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180223 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07488383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07488345 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118499 - -
          Whole Mouse Genome G4122A A_51_P287241 BC099559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 BC048675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK005888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK132924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK160153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 BC099559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 BC048675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK005888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK132924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P287241 AK160153.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180223 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09420798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09288826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09386514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09501097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09110402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09346218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09464466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09420793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09495506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09190661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09310607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09124252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09727158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09083746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09677947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09654261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09213609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09386510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09724588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09420794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28120808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09546112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10392478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392479 - -
          Affymetrix HT MG-430 PM Array Plate 1428703_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10392476 - -
          Mouse Genome 430 2.0 Array 1428703_at - -
          Mouse Genome 430 2.0 Array 1428703_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69324_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69324_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100037402.mm.1 NM_027038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037402.mm.1 NM_027038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037402.mm.1 TC1100003995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037403.mm.1 NM_001162428.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037403.mm.1 NM_001162428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037403.mm.1 NM_027038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037403.mm.1 NM_027038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037403.mm.1 TC1100003995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037401.mm.1 NM_027038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037401.mm.1 NM_027038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037401.mm.1 TC1100003995.mm.1 MTA TC
          Mouse Expression Set 430 1428703_PM_at - -
          Mouse Expression Set 430 1428703_at - -
          Murine Genome U74v2 Set 167735_r_at - -
          Murine Genome U74v2 Set 165650_f_at - -
          Murine Genome U74v2 Set 167735_PM_r_at - -
          Murine Genome U74v2 Set 165650_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1428703_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428703_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666550 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline