Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247038_m1
          See other ANKRD60 GE Assays ›
          Gene Symbol:
          Ankrd60
          Gene Name:
          ankyrin repeat domain 60
          Gene Aliases:
          1700019A24Rik, 1700030G11Rik
          Chromosome Location:
          Chr.2: 173568661 - 173578341 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247038_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd60
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199955.1 NP_001186884.1 1 - 2 348 73
          NM_027303.1 NP_081579.1 1 - 2 348 73
          GenBank mRNA AK006104.1 - 1 - 2 284 73
          BY706216.1 - 1 - 2 280 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005295.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242493 AK006546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242493 AK006546.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162028 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242493 AK006546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242493 AK006546.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162028 - -
          Whole Mouse Genome G4122A A_51_P242496 NM_027303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242496 AK006546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242493 AK006546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242493 AK006546.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139477 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100929 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02015847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02015843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02015845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01705145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01151408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01247682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01247664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01247670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01399806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01151410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01151399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01247684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01943969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01151409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02167303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21870476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02015838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10490167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490165 - -
          Affymetrix Mouse Gene 1.1 ST Array 10490164 - -
          Affymetrix Mouse Clariom S Array PSR0200045147.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045147.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045147.mm.1 NM_027303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045147.mm.1 NM_027303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045147.mm.1 TC0200005295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045143.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045143.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045143.mm.1 NM_027303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045143.mm.1 NM_027303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045143.mm.1 TC0200005295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045142.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045142.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045142.mm.1 TC0200005295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045145.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045145.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045145.mm.1 NM_027303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045145.mm.1 NM_027303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045145.mm.1 TC0200005295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045144.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045144.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045144.mm.1 NM_027303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045144.mm.1 NM_027303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045144.mm.1 TC0200005295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045146.mm.1 NM_001199955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045146.mm.1 NM_001199955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045146.mm.1 NM_027303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045146.mm.1 NM_027303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045146.mm.1 TC0200005295.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2760435 - -
          Illumina mouse-6 v1.0 expression beadchip 1940286 - -
          Illumina mouse-6 v1.0 expression beadchip 2760435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2676860 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610714 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2676860 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610714 - -
          mouse-6 v1.1 (Illumina) scl0069425.1_232-S - -
          mouse-6 v1.1 (Illumina) scl070065.1_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760435 - -
          Illumina Sentrix mouse-6.v1 bead array 1940286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070065.1_67-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069425.1_232-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760435 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940286 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2610714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2676860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline