Search
Search

Palm3| Gene Symbol: Palm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028877.1 | NP_083153.1 | 1 - 2 | 57 | 69 | |
| GenBank mRNA | AK014493.1 | - | 1 - 2 | 61 | 69 | |
| BC113196.1 | - | 1 - 2 | 60 | 69 | ||
| BC113802.1 | - | 1 - 2 | 55 | 69 | ||
| EF208034.1 | - | 1 - 2 | 57 | 69 | ||
Set Membership: |
|