Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247340_m1
          See other LCAT GE Assays ›
          Gene Symbol:
          Lcat
          Gene Name:
          lecithin cholesterol acyltransferase
          Gene Aliases:
          AI046659, D8Wsu61e
          Chromosome Location:
          Chr.8: 105939551 - 105943402 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247340_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008490.2 NP_032516.2 5 - 6 774 149
          GenBank mRNA AI574112.1 - 3 - 4 514 149
          AK004914.1 - 5 - 6 770 149
          AK149476.1 - 5 - 6 771 149
          BC028861.1 - 5 - 6 757 4212194 149
          J05154.1 - 5 - 6 754 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002843.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcat

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phospholipid metabolic process
          phosphatidylcholine biosynthetic process
          steroid metabolic process
          cholesterol metabolic process
          cholesterol transport
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          cholesterol esterification
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          reverse cholesterol transport
          response to copper ion
          response to glucocorticoid
          regulation of high-density lipoprotein particle assembly
          phosphatidylcholine-sterol O-acyltransferase activity
          phospholipase A2 activity
          O-acyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          apolipoprotein A-I binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 J05154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 NM_008490.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 AK004914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 AK149476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 J05154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 NM_008490.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 AK004914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 AK149476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533413 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 J05154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 NM_008490.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 AK004914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 AK149476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 BC028861.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 J05154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 NM_008490.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 AK004914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 AK149476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 J05154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 NM_008490.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 AK004914.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 AK149476.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 J05154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 NM_008490.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 AK004914.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 AK149476.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136894 - -
          Whole Mouse Genome G4122A A_51_P336060 J05154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 NM_008490.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 AK004914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 AK149476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 BC028861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 J05154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 NM_008490.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 AK004914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 AK149476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336060 BC028861.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06862959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06862958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06862957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06862986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06911460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06862973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581390 - -
          Affymetrix HT MG-430 PM Array Plate 1417043_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581388 - -
          Mouse Genome 430 2.0 Array 1417043_PM_at - -
          Mouse Genome 430 2.0 Array 1417043_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16816_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16816_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800021892.mm.1 NM_008490.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021892.mm.1 NM_008490.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021892.mm.1 TC0800002843.mm.1 MTA TC
          Mouse Expression Set 430 1417043_at - -
          Mouse Expression Set 430 1417043_PM_at - -
          Murine Genome U74v2 Set 161759_PM_r_at - -
          Murine Genome U74v2 Set 161759_r_at - -
          Murine Genome U74v2 Set 103023_at - -
          Murine Genome U74v2 Set 103023_PM_at - -
          Mouse Genome 430A 2.0 Array 1417043_at - -
          Mouse Genome 430A 2.0 Array 1417043_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417043_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417043_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 604131 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6400441 - -
          Illumina mouse-6 v1.0 expression beadchip 6400441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627961 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627961 - -
          mouse-6 v1.1 (Illumina) scl34385.6.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34385.6.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2798400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2798402 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2798402 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2798400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34385.6.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline