Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247365_m1
          See other HSF4 GE Assays ›
          Gene Symbol:
          Hsf4
          Gene Name:
          heat shock transcription factor 4
          Gene Aliases:
          HSF 4, HSTF 4, HSTF4, ldis1, mHSF4
          Chromosome Location:
          Chr.8: 105269748 - 105275845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256042.1 NP_001242971.1 9 - 10 1147 79
          NM_001256044.1 NP_001242973.1 8 - 9 1038 79
          NM_011939.3 NP_036069.1 9 - 10 1057 79
          NR_045689.1 9 - 10 1043 79
          GenBank mRNA AB029349.1 - 9 - 10 1079 79
          AB029350.1 - 9 - 10 984 79
          AF160966.1 - 9 - 10 1074 79
          AK148056.1 - 8 - 9 1010 79
          BC138130.1 - 9 - 10 1147 8861149 79
          BC145153.1 - 9 - 10 1043 79
          BC145154.1 - 9 - 10 1148 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001209.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsf4

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          eye development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          histone H3-K9 demethylation
          camera-type eye development
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cell development
          protein homotrimerization
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein phosphatase binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132620 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132617 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044613 AK148056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044613 NM_011939.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044613 AB029349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044613 AB029350.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76077 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139728 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044613 AK148056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044613 NM_011939.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044613 AB029349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044613 AB029350.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345867 BC145153.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345867 BC145154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345867 AF160966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345867 AK148056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345867 AK177833.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26132630 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139728 - -
          Whole Mouse Genome G4122A A_51_P345867 BC145153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345867 BC145154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345867 AF160966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345867 AK148056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P345867 AK177833.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121955 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134145 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07032609 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06887604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07032610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06887580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132620 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132621 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132617 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26132619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26132719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07354745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06887572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07354749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07032611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06887577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06806789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06887590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26132651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574660 - -
          Affymetrix HT MG-430 PM Array Plate 1425786_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574659 - -
          Mouse Genome 430 2.0 Array 1425786_a_at - -
          Mouse Genome 430 2.0 Array 1425786_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26386_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26386_at - -
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_001256042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_001256042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_001256044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_001256044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_011939.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NM_011939.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NR_045689.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 NR_045689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009507.mm.1 TC0800001209.mm.1 MTA TC
          Mouse Expression Set 430 1425786_a_at - -
          Mouse Expression Set 430 1425786_PM_a_at - -
          Murine Genome U74v2 Set 100384_PM_at - -
          Murine Genome U74v2 Set 100384_at - -
          Mouse Genome 430A 2.0 Array 1425786_a_at - -
          Mouse Genome 430A 2.0 Array 1425786_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425786_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425786_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4610273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004610273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746797 - -
          mouse-6 v1.1 (Illumina) scl33421.13.1_66-S - -
          Illumina Sentrix mouse-6.v1 bead array 4610273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33421.13.1_66-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2746797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline