Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247495_g1
          See other 4930567H12RIK GE Assays ›
          Gene Symbol:
          4930567H12Rik
          Gene Name:
          RIKEN cDNA 4930567H12 gene
          Gene Aliases:
          4930579P07Rik
          Chromosome Location:
          Chr.8: 125584638 - 125616670 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247495_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930567H12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015535.2 1 - 2 40 87
          GenBank mRNA AK016332.1 - 1 - 2 47 87
          AK131761.1 - 1 - 2 41 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001563.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2253199 AK131761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2253199 NR_015535.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162848 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01854531 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01854492 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01854474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01854517 - -
          Whole Mouse Genome G4122A A_52_P131663 AK016332.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01854492 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01854531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06894185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07351886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06698701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07328388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07047201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07047209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07376355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07322473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07351888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06865982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07330743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26252672 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800012830.mm.1 NR_015535.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012830.mm.1 TC0800001563.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800012835.mm.1 NR_015535.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012835.mm.1 TC0800001563.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 833758 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline