Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248148_m1
          See other ZFP663 GE Assays ›
          Gene Symbol:
          Zfp663
          Gene Name:
          zinc finger protein 663
          Gene Aliases:
          Gm1008
          Chromosome Location:
          Chr.2: 165351297 - 165369977 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248148_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp663
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005425.1 NP_001005425.1 4 - 5 536 97
          GenBank mRNA AK141109.1 - 5 - 6 676 97
          BC067000.1 - 4 - 5 536 5705933 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005147.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp663

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823447 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823448 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131274 BC067000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131274 NM_001005425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 BC067000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 NM_001005425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 AK141109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 BC067000.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 NM_001005425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P116973 AK141109.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75589 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106992 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131140 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131274 BC067000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131274 NM_001005425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 BC067000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 NM_001005425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 AK141109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 BC067000.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 NM_001005425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P116973 AK141109.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823433 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106992 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00551536 - -
          Whole Mouse Genome G4122A A_52_P116973 BC067000.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116973 NM_001005425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116973 AK141109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116973 BC067000.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116973 NM_001005425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116973 AK141109.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106992 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01861593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01861598 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01861591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01861599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823447 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01297382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01899480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02241269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01596265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02224650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02224649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01667730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01868981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01883045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01466848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02017935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02152603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01883044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02079431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01290659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01899491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823386 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489692 - -
          Affymetrix Mouse Gene 1.1 ST Array 10489687 - -
          Affymetrix Mouse Clariom S Array PSR0200043978.mm.1 NM_001005425.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043978.mm.1 NM_001005425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043978.mm.1 TC0200005147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043979.mm.1 NM_001005425.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043979.mm.1 NM_001005425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043979.mm.1 TC0200005147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043977.mm.1 NM_001005425.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043977.mm.1 NM_001005425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043977.mm.1 TC0200005147.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 488279 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2527809 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3093777 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3021415 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3093777 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3021415 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline