Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248389_g1
          See other TXNDC17 GE Assays ›
          Gene Symbol:
          Txndc17
          Gene Name:
          thioredoxin domain containing 17
          Gene Aliases:
          4831443O22Rik, D11Ertd672e, TRP14, Txnl5
          Chromosome Location:
          Chr.11: 72207554 - 72210487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248389_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txndc17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026559.3 NP_080835.1 3 - 4 366 69
          GenBank mRNA AJ344103.1 - 3 - 4 355 69
          AK004219.1 - 3 - 4 374 69
          AK007595.1 - 3 - 4 370 69
          AK013103.1 - 3 - 4 309 69
          AK076425.1 - 3 - 4 366 69
          AK169882.1 - 3 - 4 309 69
          BC030344.1 - 3 - 4 364 3673521 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txndc17

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          tumor necrosis factor-mediated signaling pathway
          oxidation-reduction process
          peroxidase activity
          electron carrier activity
          protein-disulfide reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27910155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27910156 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK007595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK013103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK076425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK004219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK169882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK007595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK013103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK076425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK004219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349878 AK169882.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770548 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104353 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK007595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK013103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK076425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK004219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK169882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK007595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK013103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK076425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK004219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349878 AK169882.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27910155 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104353 - -
          Whole Mouse Genome G4122A A_51_P349878 AK007595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK013103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK076425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK004219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK169882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK007595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK013103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK076425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK004219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349878 AK169882.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104075 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09292893 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27910156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27910155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27910157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09292873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27910158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378060 - -
          Affymetrix HT MG-430 PM Array Plate 1423034_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439184_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1423035_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1423036_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378059 - -
          Mouse Genome 430 2.0 Array 1423036_at - -
          Mouse Genome 430 2.0 Array 1423034_PM_at - -
          Mouse Genome 430 2.0 Array 1439184_s_at - -
          Mouse Genome 430 2.0 Array 1423035_s_at - -
          Mouse Genome 430 2.0 Array 1439184_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423036_PM_at - -
          Mouse Genome 430 2.0 Array 1423035_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423034_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52700_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52700_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008825.mm.1 NM_026559.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008825.mm.1 NM_026559.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008825.mm.1 TC1100000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008826.mm.1 NM_026559.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008826.mm.1 NM_026559.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008826.mm.1 TC1100000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008827.mm.1 NM_026559.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008827.mm.1 NM_026559.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008827.mm.1 TC1100000984.mm.1 MTA TC
          Mouse Expression Set 430 1439184_s_at - -
          Mouse Expression Set 430 1439184_PM_s_at - -
          Murine Genome U74v2 Set 94242_at - -
          Murine Genome U74v2 Set 94242_PM_at - -
          Mouse Genome 430A 2.0 Array 1439184_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1439184_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423036_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423036_at - -
          Affymetrix HT Mouse Genome 430A Array 1439184_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1439184_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423034_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423035_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423035_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423034_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 758700 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1400164 - -
          Illumina mouse-6 v1.0 expression beadchip 1190609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228991 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228991 - -
          mouse-6 v1.1 (Illumina) scl0001392.1_0-S - -
          mouse-6 v1.1 (Illumina) scl41316.5_66-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190609 - -
          Illumina Sentrix mouse-6.v1 bead array 1400164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41316.5_66-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001392.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190609 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259178 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline