Search
Search

Tmem231| Gene Symbol: Tmem231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033321.1 | NP_001028493.1 | 4 - 5 | 606 | 62 | |
| GenBank mRNA | AK030046.1 | - | 5 - 6 | 695 | 62 | |
| AK155425.1 | - | 4 - 5 | 606 | 62 | ||
| BC100402.1 | - | 4 - 5 | 590 | 30106220 | 62 | |
| BC132428.1 | - | 4 - 5 | 696 | 40130705 | 62 | |
| BC132430.1 | - | 4 - 5 | 696 | 40130707 | 62 | |
Set Membership: |
|