| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900026388.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026388.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026388.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026359.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026359.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026359.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026381.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026381.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026381.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026386.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026386.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026386.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026360.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026360.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026360.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026373.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026373.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026373.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026377.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026377.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026377.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026385.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026385.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026385.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026393.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026393.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026393.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026365.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026365.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026365.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026366.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026366.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026366.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026371.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026371.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026371.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026372.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026372.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026372.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026384.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026384.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026384.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026356.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026356.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026356.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026363.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026363.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026363.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026367.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026367.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026367.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026374.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026374.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026374.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026375.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026375.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026375.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026379.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026379.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026379.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026382.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026382.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026382.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026390.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026390.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026390.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026357.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026357.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026357.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026358.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026358.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026358.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026380.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026380.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026380.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026383.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026383.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026383.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026355.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026355.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026355.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026364.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026364.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026364.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026368.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026368.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026368.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026376.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026376.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026376.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026387.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026387.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026387.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026389.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026389.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026389.mm.1 |
TC0900003115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900026391.mm.1 |
NM_183276.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026391.mm.1 |
NM_183276.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900026391.mm.1 |
TC0900003115.mm.1 |
MTA TC |